Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012164.3 | NP_036296.2 | 8 | 1725 | ||
XM_005251910.3 | XP_005251967.1 | 8 | 1728 | |||
XM_005251913.2 | XP_005251970.1 | 8 | 1627 | |||
XM_006717049.3 | XP_006717112.1 | 8 | 1910 | |||
XM_006717051.2 | XP_006717114.1 | 7 | 1770 | |||
XM_011518518.1 | XP_011516820.1 | 8 | 1965 | |||
XM_017014615.1 | XP_016870104.1 | 7 | 1533 | |||
XM_017014616.1 | XP_016870105.1 | 8 | 1824 | |||
XM_017014617.1 | XP_016870106.1 | 8 | 1848 | |||
GenBank mRNA | AF145024.1 | - | 8 | 1631 | ||
AF176698.1 | - | 7 | 1609 | |||
AF193594.1 | - | 7 | 1436 | |||
AK022484.1 | - | 8 | 1675 | |||
AK300066.1 | - | 8 | 1556 | |||
BC018738.1 | - | 8 | 1641 | 4554961 |