Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195071.1 | NP_001182000.1 | 9 | 1560 | ||
NM_001195072.1 | NP_001182001.1 | 10 | 1646 | |||
NM_012264.4 | NP_036396.2 | 9 | 1545 | |||
XM_011530112.2 | XP_011528414.2 | 10 | 1615 | |||
XM_011530113.1 | XP_011528415.1 | 9 | 1560 | |||
XM_011530114.1 | XP_011528416.1 | 9 | 1592 | |||
XM_011530115.2 | XP_011528417.1 | 8 | 1351 | |||
XM_011530117.1 | XP_011528419.1 | 10 | 1658 | |||
XM_011530118.1 | XP_011528420.1 | 10 | 1632 | |||
XM_011530119.1 | XP_011528421.1 | 8 | 1386 | |||
XM_017028754.1 | XP_016884243.1 | 9 | 1631 | |||
XM_017028755.1 | XP_016884244.1 | 9 | 1588 | |||
XM_017028756.1 | XP_016884245.1 | 10 | 1545 | |||
XM_017028757.1 | XP_016884246.1 | 9 | 1333 | |||
XM_017028759.1 | XP_016884248.1 | 8 | 1350 | |||
GenBank mRNA | AB097053.1 | - | 9 | 1516 | ||
AK075417.1 | - | 7 | 1033 | |||
AK095109.1 | - | 1 | 1865 | |||
AL096879.1 | - | 8 | 1238 | |||
AL117649.1 | - | 1 | 121 | |||
BC012983.2 | - | 5 | 869 | |||
BC015489.1 | - | 9 | 1501 | 3896682 | ||
BX648345.1 | - | 9 | 1596 | |||
CR627445.1 | - | 1 | 2747 | |||
CR749859.1 | - | 10 | 1552 |