Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKRD17 SIRNA Assays ›
          Gene Symbol:
          ANKRD17
          Gene Name:
          ankyrin repeat domain 17
          Gene Aliases:
          GTAR, MASK2, NY-BR-16
          Chromosome Location:
          Chr.4: 73073966 - 73259142 on Build GRCh38
          UniGene ID:
          Hs.601206
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135349

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKRD17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001286771.2 NP_001273700.1 11, 12 1713
          NM_015574.1 NP_056389.1 11, 12 2095
          NM_032217.4 NP_115593.3 11, 12 2095
          NM_198889.2 NP_942592.1 11, 12 2095
          XM_005265671.3 XP_005265728.1 11, 12 2439
          XM_005265672.3 XP_005265729.1 11, 12 2439
          XM_005265673.3 XP_005265730.1 11, 12 2439
          XM_017008011.1 XP_016863500.1 11, 12 1670
          XM_017008012.1 XP_016863501.1 11, 12 1670
          XM_017008013.1 XP_016863502.1 11, 12 1670
          XM_017008014.1 XP_016863503.1 11, 12 2439
          XM_017008015.1 XP_016863504.1 11, 12 1670
          XM_017008016.1 XP_016863505.1 11, 12 2439
          XM_017008017.1 XP_016863506.1 11, 12 1670
          GenBank mRNA AB014597.2 - 11, 12 1619
          AK055209.1 - 4, 5 323
          AK299046.1 - 5, 6 595
          AK300302.1 - 10, 11 1471
          AK307581.1 - 11, 12 2095
          AL833287.1 - 8, 9 1272
          BC004173.2 - 11, 12 2076
          BC007747.2 - 9, 10 1425
          BC043394.1 - 11, 12 2064
          BX648035.1 - 11, 12 1668

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel maturation
          regulation of DNA replication
          viral process
          defense response to bacterium
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          innate immune response
          positive regulation of cell cycle
          negative regulation of smooth muscle cell differentiation
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of MDA-5 signaling pathway
          positive regulation of RIG-I signaling pathway
          chromatin binding
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0