Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMC2 SIRNA Assays ›
          Gene Symbol:
          SMC2
          Gene Name:
          structural maintenance of chromosomes 2
          Gene Aliases:
          CAP-E, CAPE, SMC-2, SMC2L1
          Chromosome Location:
          Chr.9: 104093808 - 104141419 on Build GRCh38
          UniGene ID:
          Hs.119023
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135817

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042550.1 NP_001036015.1 4, 5 799
          NM_001042551.1 NP_001036016.1 4, 5 737
          NM_001265602.1 NP_001252531.1 4, 5 753
          NM_006444.2 NP_006435.2 4, 5 728
          XM_006716933.3 XP_006716996.1 4, 5 650
          XM_011518148.2 XP_011516450.1 4, 5 613
          XM_011518149.2 XP_011516451.1 4, 5 655
          XM_011518150.2 XP_011516452.1 3, 4 517
          XM_011518151.2 XP_011516453.1 3, 4 495
          XM_011518152.2 XP_011516454.1 4, 5 679
          XM_011518153.1 XP_011516455.1 4, 5 679
          XM_017014206.1 XP_016869695.1 4, 5 799
          XM_017014207.1 XP_016869696.1 4, 5 701
          XM_017014208.1 XP_016869697.1 4, 5 741
          XM_017014209.1 XP_016869698.1 3, 4 628
          XM_017014210.1 XP_016869699.1 3, 4 518
          XM_017014211.1 XP_016869700.1 3, 4 607
          XM_017014212.1 XP_016869701.1 3, 4 474
          GenBank mRNA AF092563.1 - 4, 5 663
          AK001485.1 - 4, 5 785
          AK292599.1 - 4, 5 676
          AK307752.1 - 3, 4 495
          AL833191.1 - 4, 5 737
          AY739716.1 - 4, 5 629
          BC003396.1 - 3, 4 472
          BC017845.1 - 4, 5 667
          BC032705.1 - 4, 5 786
          BC055081.1 - 2, 3 287
          BC061906.1 - 4, 5 685
          BC071750.1 - 4, 5 799
          BC107891.1 - 4, 5 773
          BC130385.1 - 4, 5 583 40146416
          BC144163.1 - 4, 5 599 9052683

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic chromosome condensation
          meiotic chromosome condensation
          meiotic chromosome segregation
          cell division
          kinetochore organization
          protein binding
          ATP binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix