Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNF266 SIRNA Assays ›
          Gene Symbol:
          ZNF266
          Gene Name:
          zinc finger protein 266
          Gene Aliases:
          HZF1
          Chromosome Location:
          Chr.19: 9412426 - 9435578 on Build GRCh38
          UniGene ID:
          Hs.656185
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135956

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNF266
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271314.1 NP_001258243.1 11 1842
          NM_006631.3 NP_006622.2 11 1893
          XM_005259716.3 XP_005259773.1 8 1559
          XM_006722620.2 XP_006722683.1 11 1625
          XM_006722621.2 XP_006722684.1 10 1531
          XM_006722622.2 XP_006722685.1 9 1370
          XM_006722623.3 XP_006722686.1 8 1305
          XM_006722627.2 XP_006722690.1 10 2222
          XM_006722630.2 XP_006722693.1 9 1536
          XM_011527644.1 XP_011525946.1 10 1541
          XM_011527647.1 XP_011525949.1 11 2080
          XM_011527648.1 XP_011525950.1 11 2076
          XM_011527650.1 XP_011525952.1 11 1879
          XM_017026154.1 XP_016881643.1 11 1650
          XM_017026155.1 XP_016881644.1 11 1647
          XM_017026156.1 XP_016881645.1 12 1950
          XM_017026157.1 XP_016881646.1 10 2225
          XM_017026158.1 XP_016881647.1 12 1921
          XM_017026159.1 XP_016881648.1 12 1943
          XM_017026160.1 XP_016881649.1 9 2116
          XM_017026161.1 XP_016881650.1 9 1821
          XM_017026162.1 XP_016881651.1 10 1697
          XM_017026163.1 XP_016881652.1 10 1666
          XM_017026164.1 XP_016881653.1 8 1471
          XM_017026165.1 XP_016881654.1 11 1640
          XM_017026166.1 XP_016881655.1 8 1461
          XM_017026167.1 XP_016881656.1 8 1295
          XM_017026168.1 XP_016881657.1 9 2121
          XM_017026169.1 XP_016881658.1 7 1667
          XM_017026170.1 XP_016881659.1 8 1281
          XM_017026171.1 XP_016881660.1 7 1216
          XM_017026172.1 XP_016881661.1 7 1206
          XM_017026173.1 XP_016881662.1 10 1561
          GenBank mRNA AB095928.1 - 5 2678
          AK027480.1 - 1 130
          AL833471.1 - 4 798
          AL833503.1 - 11 1572
          AY376240.1 - 10 1598
          BC071628.1 - 1 149
          BC112919.1 - 9 1782 7502631
          BC126255.1 - 4 868 8991971
          BC143940.1 - 4 868
          BC146837.1 - 5 738 9056764

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA binding
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix