Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEK6 SIRNA Assays ›
          Gene Symbol:
          NEK6
          Gene Name:
          NIMA related kinase 6
          Gene Aliases:
          SID6-1512
          Chromosome Location:
          Chr.9: 124257606 - 124352442 on Build GRCh38
          UniGene ID:
          Hs.197071
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136603

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEK6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001145001.2 NP_001138473.1 10, 11 1187
          NM_001166167.1 NP_001159639.1 9, 10 969
          NM_001166168.1 NP_001159640.1 9, 10 964
          NM_001166169.1 NP_001159641.1 9, 10 973
          NM_001166170.1 NP_001159642.1 9, 10 971
          NM_001166171.1 NP_001159643.1 10, 11 1136
          NM_014397.5 NP_055212.2 9, 10 1022
          XM_005251664.1 XP_005251721.1 9, 10 1028
          XM_006716936.2 XP_006716999.1 9, 10 1033
          XM_017014217.1 XP_016869706.1 10, 11 1196
          GenBank mRNA AB026289.1 - 9, 10 953
          AF087909.1 - 9, 10 958
          AK294614.1 - 9, 10 973
          AK313071.1 - 9, 10 923
          BC000101.2 - 9, 10 968 3507262
          BC004174.2 - 10, 11 1140 2958695
          BC004209.2 - 10, 11 1140 2958695
          BC012761.2 - 9, 10 969 3640103
          CR457091.1 - 8, 9 803
          CR542222.1 - 8, 9 803

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          protein phosphorylation
          apoptotic process
          chromosome segregation
          mitotic nuclear division
          mitotic nuclear envelope disassembly
          signal transduction
          regulation of mitotic cell cycle
          peptidyl-serine phosphorylation
          regulation of mitotic metaphase/anaphase transition
          G2 DNA damage checkpoint
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          protein autophosphorylation
          spindle assembly
          regulation of cellular senescence
          magnesium ion binding
          protein serine/threonine kinase activity
          signal transducer activity
          protein binding
          ATP binding
          kinesin binding
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0