Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308160.1 | NP_001295089.1 | 4, 5 | 956 | ||
NM_014982.2 | NP_055797.2 | 4, 5 | 956 | |||
XM_005267420.4 | XP_005267477.1 | 4, 5 | 918 | |||
XM_005267421.4 | XP_005267478.1 | 4, 5 | 918 | |||
XM_005267422.4 | XP_005267479.1 | 4, 5 | 918 | |||
XM_005267423.4 | XP_005267480.1 | 4, 5 | 918 | |||
XM_005267424.3 | XP_005267481.2 | 4, 5 | 622 | |||
XM_005267425.4 | XP_005267482.1 | 4, 5 | 918 | |||
XM_005267427.4 | XP_005267484.1 | 4, 5 | 918 | |||
XM_017021096.1 | XP_016876585.1 | 4, 5 | 918 | |||
GenBank mRNA | AB023212.1 | - | 4, 5 | 955 | ||
AF233450.1 | - | 4, 5 | 956 | |||
BC140784.1 | - | 4, 5 | 622 | 9021652 |