Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308160.1 | NP_001295089.1 | 9 | 3028 | ||
NM_014982.2 | NP_055797.2 | 11 | 3361 | |||
XM_005267420.4 | XP_005267477.1 | 11 | 3323 | |||
XM_005267421.4 | XP_005267478.1 | 11 | 3320 | |||
XM_005267422.4 | XP_005267479.1 | 11 | 3308 | |||
XM_005267423.4 | XP_005267480.1 | 11 | 3308 | |||
XM_005267424.3 | XP_005267481.2 | 11 | 3027 | |||
XM_005267425.4 | XP_005267482.1 | 9 | 3005 | |||
XM_005267427.4 | XP_005267484.1 | 11 | 3323 | |||
XM_005267428.3 | XP_005267485.1 | 4 | 311 | |||
XM_006720082.3 | XP_006720145.1 | 2 | 250 | |||
XM_017021096.1 | XP_016876585.1 | 9 | 3005 | |||
XM_017021097.1 | XP_016876586.1 | 4 | 575 | |||
GenBank mRNA | AB018348.1 | - | 1 | 91 | ||
AF233450.1 | - | 11 | 3361 | |||
BC140784.1 | - | 9 | 2694 | 9021652 |