Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304815.1 | NP_001291744.1 | 19, 20 | 7116 | ||
NM_015125.4 | NP_055940.3 | 18, 19 | 4361 | |||
XM_005258673.2 | XP_005258730.1 | 18, 19 | 4447 | |||
XM_005258674.2 | XP_005258731.1 | 18, 19 | 4447 | |||
XM_005258675.2 | XP_005258732.1 | 18, 19 | 4444 | |||
XM_011526660.2 | XP_011524962.1 | 19, 20 | 7158 | |||
XM_011526661.2 | XP_011524963.1 | 19, 20 | 7155 | |||
XM_011526662.2 | XP_011524964.1 | 19, 20 | 7155 | |||
XM_011526663.2 | XP_011524965.1 | 19, 20 | 7155 | |||
XM_011526664.2 | XP_011524966.1 | 19, 20 | 7152 | |||
GenBank mRNA | AB002304.1 | - | 15, 16 | 3852 | ||
AF363689.1 | - | 18, 19 | 4361 | |||
AK025939.1 | - | 3, 4 | 426 |