Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other R3HDM1 SIRNA Assays ›
          Gene Symbol:
          R3HDM1
          Gene Name:
          R3H domain containing 1
          Gene Aliases:
          R3HDM
          Chromosome Location:
          Chr.2: 135531429 - 135725270 on Build GRCh38
          UniGene ID:
          Hs.412462
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136914

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: R3HDM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282798.1 NP_001269727.1 4, 5 647
          NM_015361.3 NP_056176.2 4, 5 642
          XM_005263626.3 XP_005263683.1 4, 5 631
          XM_005263627.3 XP_005263684.1 4, 5 631
          XM_005263628.3 XP_005263685.1 4, 5 631
          XM_005263629.3 XP_005263686.1 4, 5 631
          XM_006712389.2 XP_006712452.1 4, 5 631
          XM_011510870.1 XP_011509172.1 4, 5 631
          XM_011510871.1 XP_011509173.1 4, 5 647
          XM_011510873.2 XP_011509175.1 4, 5 631
          XM_011510876.2 XP_011509178.1 4, 5 631
          XM_011510877.2 XP_011509179.1 4, 5 631
          XM_011510878.2 XP_011509180.1 4, 5 631
          XM_011510879.2 XP_011509181.1 4, 5 631
          XM_011510881.1 XP_011509183.1 4, 5 631
          XM_017003718.1 XP_016859207.1 4, 5 647
          XM_017003720.1 XP_016859209.1 4, 5 647
          XM_017003721.1 XP_016859210.1 4, 5 647
          XM_017003725.1 XP_016859214.1 5, 6 723
          XM_017003726.1 XP_016859215.1 4, 5 631
          XM_017003728.1 XP_016859217.1 4, 5 631
          XM_017003729.1 XP_016859218.1 4, 5 647
          XM_017003731.1 XP_016859220.1 5, 6 744
          XM_017003732.1 XP_016859221.1 5, 6 722
          XM_017003733.1 XP_016859222.1 4, 5 633
          XM_017003734.1 XP_016859223.1 4, 5 631
          XM_017003736.1 XP_016859225.1 4, 5 628
          GenBank mRNA AK290015.1 - 4, 5 533
          BC041093.1 - 4, 5 584 5556247

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001134867, XM_008769462, XM_008769461, XM_008769460, XM_008769459, and others.
          This siRNA also targets the following transcripts in Mouse: NM_181750, AK051765, BC144899, BC137772, BC049181.

          Gene Ontology Categories:

          Process(es)

          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0