Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083585.2 | NP_001077054.1 | 8, 9 | 1330 | ||
NM_001291581.1 | NP_001278510.1 | 7, 8 | 1201 | |||
NM_004703.5 | NP_004694.2 | 8, 9 | 1330 | |||
XM_011524060.1 | XP_011522362.1 | 9, 10 | 1396 | |||
XM_017025294.1 | XP_016880783.1 | 7, 8 | 923 | |||
XM_017025295.1 | XP_016880784.1 | 6, 7 | 857 | |||
GenBank mRNA | AF098638.1 | - | 8, 9 | 1106 | ||
AK297535.1 | - | 7, 8 | 1190 | |||
AK310932.1 | - | 7, 8 | 1142 | |||
AK314183.1 | - | 8, 9 | 1321 | |||
BC032435.1 | - | 8, 9 | 1284 | |||
BC041700.1 | - | 8, 9 | 1296 | 6046320 | ||
X77723.1 | - | 8, 9 | 1179 | |||
X91141.1 | - | 8, 9 | 1294 |