Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004870.3 | NP_004861.2 | 4, 5 | 612 | ||
NR_024603.1 | 4, 5 | 607 | ||||
XM_006721598.2 | XP_006721661.1 | 4, 5 | 411 | |||
XM_011524081.2 | XP_011522383.1 | 4, 5 | 443 | |||
GenBank mRNA | AF038961.1 | - | 4, 5 | 412 | ||
AF059752.1 | - | 4, 5 | 452 | |||
AK075299.1 | - | 4, 5 | 411 | |||
AK225401.1 | - | 4, 5 | 411 | |||
AK225432.1 | - | 4, 5 | 407 | |||
AK297001.1 | - | 4, 5 | 407 | |||
AK300140.1 | - | 4, 5 | 398 | |||
AK314527.1 | - | 4, 5 | 407 | |||
BC001898.1 | - | 4, 5 | 423 | 3536216 | ||
DQ499597.1 | - | 4, 5 | 396 | |||
KU178478.1 | - | 4, 5 | 396 |