Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MELK SIRNA Assays ›
          Gene Symbol:
          MELK
          Gene Name:
          maternal embryonic leucine zipper kinase
          Gene Aliases:
          HPK38
          Chromosome Location:
          Chr.9: 36572862 - 36677683 on Build GRCh38
          UniGene ID:
          Hs.184339
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138433

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MELK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256685.1 NP_001243614.1 13, 14 1477
          NM_001256687.1 NP_001243616.1 13, 14 1456
          NM_001256688.1 NP_001243617.1 12, 13 1387
          NM_001256689.1 NP_001243618.1 13, 14 1514
          NM_001256690.1 NP_001243619.1 12, 13 1397
          NM_001256691.1 NP_001243620.1 12, 13 1370
          NM_001256692.1 NP_001243621.1 11, 12 1274
          NM_001256693.1 NP_001243622.1 12, 13 1414
          NM_014791.3 NP_055606.1 14, 15 1600
          NR_046337.1 14, 15 1637
          XM_011518076.2 XP_011516378.1 14, 15 1779
          XM_011518077.1 XP_011516379.1 15, 16 1675
          XM_011518078.2 XP_011516380.1 15, 16 1862
          XM_011518079.1 XP_011516381.1 14, 15 1552
          XM_011518080.1 XP_011516382.1 13, 14 1480
          XM_011518081.2 XP_011516383.1 14, 15 1585
          XM_011518082.2 XP_011516384.1 13, 14 1693
          XM_011518083.2 XP_011516385.1 14, 15 1776
          XM_011518084.2 XP_011516386.1 13, 14 1388
          XM_011518085.1 XP_011516387.1 12, 13 1384
          GenBank mRNA AB183427.1 - 13, 14 1465
          AB183428.1 - 12, 13 1350
          AB267395.1 - 2, 3 328
          AK222811.1 - 14, 15 1599
          AK293284.1 - 12, 13 1352
          AK293423.1 - 12, 13 1335
          AK293447.1 - 12, 13 1354
          AK296820.1 - 14, 15 1636
          AK299164.1 - 13, 14 1429
          AK300821.1 - 13, 14 1468
          AK301131.1 - 12, 13 1346
          AK302374.1 - 11, 12 1241
          BC014039.2 - 14, 15 1554 4547136
          D79997.1 - 14, 15 1586

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          apoptotic process
          cell proliferation
          intrinsic apoptotic signaling pathway in response to oxidative stress
          peptidyl-tyrosine phosphorylation
          hemopoiesis
          intracellular signal transduction
          positive regulation of apoptotic process
          protein autophosphorylation
          neural precursor cell proliferation
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          calcium ion binding
          protein binding
          ATP binding
          lipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix