Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003395.2 | NP_001003395.1 | 7 | 1378 | ||
NM_001003396.2 | NP_001003396.1 | 6 | 1215 | |||
NM_001003397.2 | NP_001003397.1 | 5 | 1176 | |||
NM_001292026.2 | NP_001278955.1 | 5 | 1278 | |||
NM_001300994.2 | NP_001287923.1 | 6 | 1237 | |||
NM_001318903.1 | NP_001305832.1 | 8 | 1291 | |||
NM_001318907.1 | NP_001305836.1 | 5 | 1073 | |||
NM_003287.3 | NP_003278.1 | 7 | 1276 | |||
XM_005267122.2 | XP_005267179.1 | 7 | 1057 | |||
XM_006715557.3 | XP_006715620.1 | 7 | 1544 | |||
XM_017011239.1 | XP_016866728.1 | 6 | 1317 | |||
XM_017011240.1 | XP_016866729.1 | 5 | 956 | |||
GenBank mRNA | AF004427.1 | - | 5 | 993 | ||
AK090516.1 | - | 1 | 2390 | |||
AK291866.1 | - | 5 | 1140 | |||
AK291887.1 | - | 7 | 1071 | |||
BC002375.2 | - | 6 | 1105 | 2822877 | ||
U44427.1 | - | 7 | 1233 | |||
U44428.1 | - | 4 | 739 | |||
U44429.1 | - | 2 | 622 |