Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045104.1 | 2 | 403 | |||
NR_045105.2 | 2 | 657 | ||||
NR_045106.2 | 2 | 564 | ||||
NR_045901.1 | 2 | 403 | ||||
XM_006720708.3 | XP_006720771.1 | 2 | 339 | |||
GenBank mRNA | AK027475.1 | - | 2 | 245 | ||
AK296134.1 | - | 2 | 630 | |||
AK301222.1 | - | 2 | 356 | |||
AK310857.1 | - | 2 | 249 | |||
AL136662.1 | - | 2 | 485 | |||
BC007991.2 | - | 2 | 230 | 3140504 | ||
BC037823.1 | - | 1 | 955 | |||
DC313962.1 | - | 2 | 548 |