Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB41L2 SIRNA Assays ›
          Gene Symbol:
          EPB41L2
          Gene Name:
          erythrocyte membrane protein band 4.1 like 2
          Gene Aliases:
          4.1-G, 4.1G
          Chromosome Location:
          Chr.6: 130829111 - 131063322 on Build GRCh38
          UniGene ID:
          Hs.486470
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 145899

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPB41L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135554.1 NP_001129026.1 9 1544
          NM_001135555.3 NP_001129027.1 9 1557
          NM_001199388.2 NP_001186317.1 9 1557
          NM_001252660.1 NP_001239589.1 9 1524
          NM_001431.3 NP_001422.1 9 1544
          XM_006715362.2 XP_006715425.1 9 1575
          XM_011535524.1 XP_011533826.1 9 1544
          XM_011535525.1 XP_011533827.1 9 1544
          XM_011535527.1 XP_011533829.1 9 1544
          XM_011535528.1 XP_011533830.1 9 1544
          XM_011535529.1 XP_011533831.1 9 1544
          XM_011535530.1 XP_011533832.1 9 1544
          XM_011535534.1 XP_011533836.1 9 1544
          XM_011535536.1 XP_011533838.1 9 1544
          XM_017010349.1 XP_016865838.1 9 1572
          XM_017010350.1 XP_016865839.1 9 1506
          XM_017010351.1 XP_016865840.1 9 1568
          XM_017010352.1 XP_016865841.1 9 1410
          XM_017010353.1 XP_016865842.1 9 1544
          XM_017010354.1 XP_016865843.1 9 1544
          XM_017010355.1 XP_016865844.1 9 1572
          XM_017010356.1 XP_016865845.1 9 1544
          XM_017010357.1 XP_016865846.1 9 1506
          XM_017010358.1 XP_016865847.1 9 1544
          XM_017010359.1 XP_016865848.1 9 1572
          XM_017010360.1 XP_016865849.1 9 1410
          XM_017010361.1 XP_016865850.1 9 1572
          XM_017010362.1 XP_016865851.1 9 1506
          XM_017010363.1 XP_016865852.1 9 1534
          XM_017010364.1 XP_016865853.1 7 793
          GenBank mRNA AB209522.1 - 9 1455
          AF027299.1 - 9 1406
          AK295124.1 - 9 1486
          AK295958.1 - 1 16
          AK307764.1 - 9 1577
          AY047584.1 - 8 1370
          CR749262.1 - 9 1486
          CR936793.1 - 9 1524

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cortical actin cytoskeleton organization
          actin binding
          structural molecule activity
          spectrin binding
          PH domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix