Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145513.1 | NP_001138985.1 | 3 | 379 | ||
NM_001145514.1 | NP_001138986.1 | 3 | 311 | |||
NM_001145515.1 | NP_001138987.1 | 2 | 211 | |||
NM_014766.4 | NP_055581.3 | 3 | 371 | |||
GenBank mRNA | AB071705.1 | - | 3 | 261 | ||
AK289514.1 | - | 2 | 406 | |||
AK293462.1 | - | 3 | 323 | |||
AK295172.1 | - | 3 | 311 | |||
AK295713.1 | - | 2 | 114 | |||
BC040492.1 | - | 3 | 323 | 5314711 | ||
D83777.2 | - | 3 | 316 | |||
DC307846.1 | - | 3 | 380 | |||
DC370097.1 | - | 3 | 371 |