Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300816.2 | NP_001287745.1 | 5 | 1086 | ||
NM_001300817.2 | NP_001287746.1 | 5 | 1068 | |||
NM_001300818.2 | NP_001287747.1 | 4 | 1044 | |||
NM_001300819.2 | NP_001287748.1 | 3 | 978 | |||
NM_001300822.2 | NP_001287751.1 | 5 | 1114 | |||
NM_001308127.1 | NP_001295056.1 | 5 | 1116 | |||
NM_001308128.1 | NP_001295057.1 | 3 | 858 | |||
NM_001308129.1 | NP_001295058.1 | 2 | 816 | |||
NM_001308130.1 | NP_001295059.1 | 4 | 1048 | |||
NM_017665.3 | NP_060135.1 | 4 | 1020 | |||
NR_125337.2 | 4 | 1024 | ||||
NR_131766.1 | 5 | 1169 | ||||
NR_131767.1 | 4 | 1052 | ||||
XM_011543502.1 | XP_011541804.1 | 4 | 1073 | |||
XM_011543503.2 | XP_011541805.1 | 5 | 1161 | |||
GenBank mRNA | AK000101.1 | - | 4 | 964 | ||
AK122970.1 | - | 5 | 1013 | |||
BC005211.1 | - | 4 | 977 | 3961163 | ||
BC022443.1 | - | 5 | 1027 | 4285562 |