Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANK3 SIRNA Assays ›
          Gene Symbol:
          ANK3
          Gene Name:
          ankyrin 3, node of Ranvier (ankyrin G)
          Gene Aliases:
          ANKYRIN-G, MRT37
          Chromosome Location:
          Chr.10: 60026298 - 60733526 on Build GRCh38
          UniGene ID:
          Hs.499725
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147781

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001149.3 NP_001140.2 7 1563
          NM_001204403.1 NP_001191332.1 30 3701
          NM_001204404.1 NP_001191333.1 30 3944
          NM_001320874.1 NP_001307803.1 29 3958
          NM_020987.4 NP_066267.2 29 3958
          XM_005269715.3 XP_005269772.1 30 3845
          XM_006717796.3 XP_006717859.1 31 3857
          XM_006717802.3 XP_006717865.1 31 3857
          XM_011539708.2 XP_011538010.1 31 3857
          XM_011539709.2 XP_011538011.1 31 3857
          XM_017016110.1 XP_016871599.1 31 3857
          XM_017016111.1 XP_016871600.1 30 3845
          XM_017016112.1 XP_016871601.1 31 3842
          XM_017016113.1 XP_016871602.1 31 3857
          XM_017016114.1 XP_016871603.1 31 3956
          XM_017016115.1 XP_016871604.1 31 3686
          XM_017016116.1 XP_016871605.1 31 3857
          XM_017016117.1 XP_016871606.1 31 3857
          XM_017016118.1 XP_016871607.1 31 3857
          XM_017016119.1 XP_016871608.1 31 3857
          XM_017016120.1 XP_016871609.1 31 3857
          XM_017016121.1 XP_016871610.1 31 3857
          XM_017016122.1 XP_016871611.1 29 3791
          XM_017016123.1 XP_016871612.1 31 3857
          XM_017016124.1 XP_016871613.1 31 3857
          XM_017016125.1 XP_016871614.1 30 3830
          XM_017016126.1 XP_016871615.1 29 3725
          XM_017016127.1 XP_016871616.1 29 3725
          XM_017016128.1 XP_016871617.1 31 3857
          XM_017016129.1 XP_016871618.1 31 3857
          XM_017016130.1 XP_016871619.1 29 3791
          XM_017016131.1 XP_016871620.1 29 3776
          XM_017016132.1 XP_016871621.1 29 3791
          XM_017016133.1 XP_016871622.1 29 3890
          XM_017016134.1 XP_016871623.1 29 3725
          XM_017016135.1 XP_016871624.1 29 3890
          XM_017016136.1 XP_016871625.1 31 3857
          XM_017016137.1 XP_016871626.1 31 3857
          XM_017016138.1 XP_016871627.1 31 3857
          XM_017016139.1 XP_016871628.1 29 3791
          XM_017016140.1 XP_016871629.1 29 3791
          XM_017016141.1 XP_016871630.1 29 3725
          XM_017016142.1 XP_016871631.1 28 3713
          XM_017016143.1 XP_016871632.1 29 3725
          XM_017016144.1 XP_016871633.1 29 3725
          XM_017016145.1 XP_016871634.1 28 3713
          XM_017016146.1 XP_016871635.1 28 3713
          XM_017016147.1 XP_016871636.1 28 3713
          XM_017016148.1 XP_016871637.1 29 3607
          XM_017016149.1 XP_016871638.1 28 3812
          XM_017016150.1 XP_016871639.1 29 3791
          GenBank mRNA AB209308.1 - 30 3846
          AK092527.1 - 7 1568
          AK126851.1 - 11 1646
          AK292927.1 - 7 1449
          AK295661.1 - 30 3651
          AL136710.1 - 7 1222
          BX537917.1 - 30 3572
          BX648574.1 - 30 3666
          U13616.1 - 29 3704

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          ER to Golgi vesicle-mediated transport
          plasma membrane organization
          cytoskeletal anchoring at plasma membrane
          signal transduction
          axonogenesis
          axon guidance
          neuromuscular junction development
          positive regulation of gene expression
          positive regulation of cell communication by electrical coupling
          positive regulation of sodium ion transport
          magnesium ion homeostasis
          neuronal action potential
          positive regulation of homotypic cell-cell adhesion
          Golgi to plasma membrane protein transport
          regulation of potassium ion transport
          establishment of protein localization
          positive regulation of membrane potential
          cellular response to magnesium ion
          membrane assembly
          protein localization to plasma membrane
          maintenance of protein location in plasma membrane
          protein targeting to plasma membrane
          positive regulation of protein targeting to membrane
          positive regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of delayed rectifier potassium channel activity
          positive regulation of sodium ion transmembrane transporter activity
          positive regulation of cation channel activity
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          cytoskeletal adaptor activity
          spectrin binding
          protein binding, bridging
          ion channel binding
          cadherin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix