Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 10 | 1749 | ||
NM_001127454.1 | NP_001120926.1 | 9 | 1764 | |||
NM_004403.2 | NP_004394.1 | 10 | 1994 | |||
XM_017011802.1 | XP_016867291.1 | 9 | 1520 | |||
GenBank mRNA | AF007790.2 | - | 9 | 1648 | ||
AF073308.1 | - | 10 | 1718 | |||
AF075171.1 | - | 2 | 1403 | |||
AF131765.1 | - | 2 | 1405 | |||
AJ270686.1 | - | 1 | 19 | |||
AK094714.1 | - | 12 | 2322 | |||
BC099911.1 | - | 10 | 1710 | 5287362 | ||
BC125065.1 | - | 10 | 1716 | 40117291 | ||
BC125066.1 | - | 9 | 1486 | |||
BX647389.1 | - | 2 | 3681 |