Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032423.2 | NP_115799.2 | 7 | 846 | ||
XM_005259323.1 | XP_005259380.1 | 4 | 429 | |||
XM_006723418.2 | XP_006723481.1 | 9 | 1094 | |||
XM_011527392.1 | XP_011525694.1 | 9 | 1090 | |||
XM_011527394.2 | XP_011525696.1 | 4 | 962 | |||
XM_017027360.1 | XP_016882849.1 | 10 | 1211 | |||
XM_017027361.1 | XP_016882850.1 | 10 | 1207 | |||
XM_017027362.1 | XP_016882851.1 | 8 | 1006 | |||
XM_017027363.1 | XP_016882852.1 | 9 | 1000 | |||
XM_017027364.1 | XP_016882853.1 | 8 | 955 | |||
XM_017027365.1 | XP_016882854.1 | 4 | 505 | |||
GenBank mRNA | AK056552.1 | - | 8 | 1078 | ||
AL832630.2 | - | 1 | 1669 | |||
AL833325.1 | - | 7 | 846 | |||
BC101712.1 | - | 4 | 462 | 8069218 | ||
BC111950.1 | - | 4 | 474 | 8327418 | ||
BX640670.1 | - | 3 | 3246 |