Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008891.2 | NP_032917.2 | 2 | 255 | ||
GenBank mRNA | AK009055.1 | - | 2 | 210 | ||
AK028219.1 | - | 2 | 196 | |||
AK030978.1 | - | 2 | 184 | |||
AK050669.1 | - | 2 | 201 | |||
AK051640.1 | - | 2 | 253 | |||
AK157587.1 | - | 2 | 199 | |||
AK160616.1 | - | 2 | 190 | |||
AK161109.1 | - | 2 | 195 | |||
BC132571.1 | - | 2 | 189 | 40130848 | ||
BC138014.1 | - | 2 | 189 | 8861033 | ||
BY775218.1 | - | 2 | 256 | |||
Y08701.1 | - | 2 | 189 |