Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166480.1 | NP_001159952.1 | 4 | 522 | ||
NM_009279.3 | NP_033305.1 | 4 | 404 | |||
GenBank mRNA | AK003044.1 | - | 4 | 384 | ||
AK007921.1 | - | 4 | 360 | |||
AK013723.1 | - | 4 | 366 | |||
AK160089.1 | - | 4 | 370 | |||
AK191259.1 | - | 2 | 104 | |||
BC008172.1 | - | 4 | 325 | 3481948 | ||
BC012971.1 | - | 2 | 2061 | |||
BC019419.1 | - | 3 | 1855 | |||
BI989805.1 | - | 4 | 522 | |||
BQ747160.1 | - | 4 | 307 | |||
X90582.1 | - | 4 | 485 |