Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011403.2 | NP_035533.1 | 13 | 1883 | ||
GenBank mRNA | AK137542.1 | - | 13 | 1779 | ||
AK157181.1 | - | 13 | 1852 | |||
AK157833.1 | - | 13 | 1741 | |||
AK157861.1 | - | 13 | 1753 | |||
AK158143.1 | - | 13 | 1884 | |||
AK158184.1 | - | 13 | 1849 | |||
AK158243.1 | - | 13 | 1753 | |||
AK158453.1 | - | 13 | 1728 | |||
AK162616.1 | - | 13 | 1728 | |||
AK165195.1 | - | 13 | 1740 | |||
AK165267.1 | - | 13 | 1747 | |||
AK171662.1 | - | 1779 | ||||
AK171808.1 | - | 13 | 1717 | |||
AK171845.1 | - | 1779 | ||||
AK172103.1 | - | 13 | 1740 | |||
AK172316.1 | - | 13 | 1728 | |||
AK172395.1 | - | 13 | 1779 | |||
AK172456.1 | - | 13 | 1763 | |||
AK172569.1 | - | 13 | 1728 | |||
AY296129.1 | - | 13 | 1716 | |||
BC052419.1 | - | 13 | 1723 | 5702599 | ||
BC053429.1 | - | 13 | 1714 | 6506270 | ||
M29379.1 | - | 13 | 1710 | |||
X02677.1 | - | 13 | 1716 | |||
X03917.1 | - | 11 | 1560 |