Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC8A1 SIRNA Assays ›
          Gene Symbol:
          Slc8a1
          Gene Name:
          solute carrier family 8 (sodium/calcium exchanger), member 1
          Gene Aliases:
          AI852629, AV344025, D930008O12Rik, Ncx1
          Chromosome Location:
          Chr.17: 81373105 - 81738387 on Build GRCm38
          UniGene ID:
          Mm.265834
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 151755

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Slc8a1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001112798.2 NP_001106269.1 2 400
          NM_001286684.1 NP_001273613.1 2 400
          NM_011406.3 NP_035536.2 2 400
          GenBank mRNA AF004666.1 - 2 395
          BB660129.1 - 2 400
          BC079673.1 - 1 291 30532156
          CJ140501.1 - 2 402
          DQ388998.1 - 1 290
          U70033.1 - 2 390

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001270777, NM_001270776, NM_001270772, NM_001270775, NM_001270779, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          embryonic placenta development
          regulation of the force of heart contraction
          regulation of heart rate
          regulation of sodium ion transport
          heart morphogenesis
          transport
          ion transport
          sodium ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          cellular sodium ion homeostasis
          cell communication
          positive regulation of cytosolic calcium ion concentration
          post-embryonic development
          positive regulation of fibroblast migration
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          vascular smooth muscle contraction
          positive regulation of bone mineralization
          response to ATP
          cellular response to reactive oxygen species
          embryonic heart tube development
          sodium ion transmembrane transport
          response to muscle stretch
          relaxation of smooth muscle
          muscle fiber development
          negative regulation of cytosolic calcium ion concentration
          regulation of calcium ion transport
          cardiac muscle cell development
          calcium ion homeostasis
          transmembrane transport
          cardiac muscle contraction
          calcium ion transport into cytosol
          calcium ion import
          calcium ion transmembrane transport
          cellular response to caffeine
          cellular response to cAMP
          sodium ion export
          regulation of cardiac muscle cell membrane potential
          cell communication by electrical coupling involved in cardiac conduction
          sodium ion import
          positive regulation of the force of heart contraction
          calcium ion export
          calcium:sodium antiporter activity
          protein binding
          calmodulin binding
          cytoskeletal protein binding
          antiporter activity
          ankyrin binding
          ion channel binding
          metal ion binding
          calcium:sodium antiporter activity involved in regulation of cardiac muscle cell membrane potential

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix