Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013663.5 | NP_038691.1 | 5 | 547 | ||
GenBank mRNA | AK009895.1 | - | 5 | 535 | ||
AK011657.1 | - | 5 | 535 | |||
AK078891.1 | - | 5 | 535 | |||
AK088122.1 | - | 5 | 533 | |||
AK134177.1 | - | 5 | 535 | |||
AK135298.1 | - | 5 | 535 | |||
AK141440.1 | - | 3 | 3078 | |||
AK145089.1 | - | 5 | 536 | |||
AK145600.1 | - | 5 | 535 | |||
AK145715.1 | - | 5 | 535 | |||
AK146010.1 | - | 5 | 535 | |||
AK146253.1 | - | 5 | 533 | |||
AK146610.1 | - | 5 | 535 | |||
AK150912.1 | - | 5 | 535 | |||
AK152411.1 | - | 6 | 991 | |||
AK152704.1 | - | 5 | 536 | |||
AK152779.1 | - | 6 | 991 | |||
AK153073.1 | - | 6 | 991 | |||
AK153209.1 | - | 5 | 535 | |||
AK154046.1 | - | 5 | 535 | |||
BC068111.1 | - | 5 | 527 | 5710934 | ||
BC071196.1 | - | 5 | 509 | 30441615 | ||
BC083316.1 | - | 5 | 519 | 6818630 | ||
X53824.1 | - | 5 | 480 |