Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009031.3 | NP_033057.3 | 6 | 986 | ||
GenBank mRNA | AK076016.1 | - | 6 | 911 | ||
AK078349.1 | - | 6 | 899 | |||
AK082892.1 | - | 6 | 898 | |||
AK132101.1 | - | 6 | 897 | |||
AK135779.1 | - | 6 | 900 | |||
AK135869.1 | - | 5 | 595 | |||
AK135875.1 | - | 5 | 595 | |||
AK135956.1 | - | 6 | 900 | |||
AK136110.1 | - | 6 | 898 | |||
AK145531.1 | - | 6 | 897 | |||
AK145651.1 | - | 6 | 880 | |||
AK146014.1 | - | 6 | 898 | |||
AK146439.1 | - | 6 | 957 | |||
AK146904.1 | - | 6 | 899 | |||
AK146967.1 | - | 6 | 898 | |||
AK147062.1 | - | 6 | 957 | |||
AK148852.1 | - | 6 | 926 | |||
AK153913.1 | - | 6 | 943 | |||
AK160023.1 | - | 6 | 898 | |||
BC003785.1 | - | 6 | 915 | 3600013 | ||
U35142.1 | - | 6 | 925 |