Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130529.2 | NP_001124001.1 | 10 | 2545 | ||
NM_001291014.1 | NP_001277943.1 | 10 | 2299 | |||
NM_172941.4 | NP_766529.3 | 10 | 2592 | |||
GenBank mRNA | AK051711.1 | - | 10 | 1881 | ||
AK155196.1 | - | 10 | 2122 | |||
AK155490.1 | - | 10 | 2174 | |||
AK157002.1 | - | 4 | 254 | |||
AK170924.1 | - | 10 | 2122 | |||
AK170989.1 | - | 10 | 2127 | |||
AK171190.1 | - | 10 | 2174 | |||
AY302242.1 | - | 9 | 2073 | |||
BC040205.1 | - | 9 | 2077 | 4947290 |