Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253831.1 | NP_001240760.1 | 4 | 501 | ||
NM_001253834.1 | NP_001240763.1 | 5 | 592 | |||
NM_001253836.1 | NP_001240765.1 | 4 | 524 | |||
NM_175025.4 | NP_778190.3 | 5 | 707 | |||
GenBank mRNA | AJ551270.1 | - | 3 | 289 | ||
AK007226.1 | - | 6 | 493 | |||
AK028393.1 | - | 4 | 470 | |||
AK133703.1 | - | 4 | 502 | |||
AK136062.1 | - | 592 | ||||
AK159918.1 | - | 5 | 658 | |||
AK163049.1 | - | 5 | 553 | |||
AK173154.1 | - | 3 | 280 | |||
BC043091.1 | - | 5 | 673 | 6405251 | ||
BC043706.1 | - | 5 | 654 |