Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDH18 SIRNA Assays ›
          Gene Symbol:
          CDH18
          Gene Name:
          cadherin 18
          Gene Aliases:
          CDH14, CDH14L, CDH24
          Chromosome Location:
          Chr.5: 19472930 - 20575902 on Build GRCh38
          UniGene ID:
          Hs.317632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15484

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDH18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001167667.1 NP_001161139.1 3 638
          NM_001291956.1 NP_001278885.1 5 1095
          NM_001291957.1 NP_001278886.1 3 616
          NM_004934.3 NP_004925.1 3 638
          XM_005248228.3 XP_005248285.1 3 847
          XM_006714435.3 XP_006714498.1 2 454
          XM_011513930.2 XP_011512232.1 3 725
          XM_017008924.1 XP_016864413.1 4 966
          XM_017008925.1 XP_016864414.1 4 962
          XM_017008926.1 XP_016864415.1 4 823
          XM_017008927.1 XP_016864416.1 4 752
          XM_017008928.1 XP_016864417.1 5 1010
          XM_017008929.1 XP_016864418.1 2 490
          XM_017008930.1 XP_016864419.1 2 381
          XM_017008931.1 XP_016864420.1 3 634
          XM_017008932.1 XP_016864421.1 4 685
          XM_017008933.1 XP_016864422.1 3 725
          XM_017008934.1 XP_016864423.1 3 863
          XM_017008935.1 XP_016864424.1 4 966
          XM_017008936.1 XP_016864425.1 3 847
          XM_017008937.1 XP_016864426.1 3 606
          XM_017008938.1 XP_016864427.1 3 570
          XM_017008939.1 XP_016864428.1 4 943
          XM_017008940.1 XP_016864429.1 3 831
          XM_017008941.1 XP_016864430.1 3 590
          GenBank mRNA AK289547.1 - 5 1093
          AK295087.1 - 3 579
          AK308745.1 - 3 616
          BC031051.1 - 3 594 5278717
          BI549773.1 - 3 598
          U59325.1 - 2 417

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          adherens junction organization
          calcium ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0