Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130150.1 | NP_001123622.1 | 10 | 960 | ||
NM_001130151.1 | NP_001123623.1 | 10 | 960 | |||
NM_001130152.1 | NP_001123624.1 | 10 | 969 | |||
NM_001130153.1 | NP_001123625.1 | 10 | 960 | |||
NM_008488.2 | NP_032514.1 | 10 | 960 | |||
GenBank mRNA | AK043027.1 | - | 10 | 924 | ||
AK051518.1 | - | 10 | 910 | |||
AK131181.1 | - | 10 | 857 | |||
AK157056.1 | - | 10 | 896 | |||
AK172354.1 | - | 10 | 896 | |||
BC012488.1 | - | 10 | 920 | 3154558 | ||
CA320228.1 | - | 3 | 192 | |||
U58203.1 | - | 10 | 890 |