Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171187.1 | NP_001164658.1 | 3 | 1637 | ||
NM_010762.5 | NP_034892.1 | 4 | 1805 | |||
GenBank mRNA | AK004639.1 | - | 4 | 1392 | ||
AK158653.1 | - | 4 | 1806 | |||
AK169172.1 | - | 4 | 1329 | |||
BC006826.1 | - | 4 | 1302 | 3597561 |