Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080773.2 | NP_001074242.1 | 13 | 1592 | ||
NM_001286662.1 | NP_001273591.1 | 13 | 1510 | |||
NM_011062.4 | NP_035192.2 | 13 | 1592 | |||
GenBank mRNA | AF079535.1 | - | 13 | 1549 | ||
AF086625.1 | - | 13 | 1558 | |||
AF126294.1 | - | 13 | 1533 | |||
AK147149.1 | - | 13 | 1584 | |||
AK147835.1 | - | 13 | 1567 | |||
AK149162.1 | - | 13 | 1526 | |||
AK162677.1 | - | 13 | 1552 | |||
AK165564.1 | - | 13 | 1530 | |||
AY062008.1 | - | 13 | 1488 | |||
BC146001.1 | - | 13 | 1529 | 40131304 | ||
BC146003.1 | - | 13 | 1529 | 40131306 |