Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB560752.1 | - | 5 | 1054 | ||
AB560753.1 | - | 4 | 917 | |||
AB560754.1 | - | 4 | 917 | |||
AB560755.1 | - | 4 | 917 | |||
AB560756.1 | - | 4 | 917 | |||
AF128220.1 | - | 1 | 44 | |||
AF128221.1 | - | 1 | 44 | |||
AK036627.1 | - | 5 | 1028 | |||
AK039911.1 | - | 8 | 1531 | |||
AK041525.1 | - | 6 | 1176 | |||
AK054182.1 | - | 5 | 1059 | |||
AK077236.1 | - | 5 | 1033 | |||
AK087638.1 | - | 5 | 1008 | |||
AK139745.1 | - | 6 | 1588 | |||
EU791538.1 | - | 5 | 972 | |||
EU791540.1 | - | 5 | 1097 | |||
M38651.1 | - | 5 | 1036 |