Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079908.2 | NP_001073377.1 | 16 | 2903 | ||
NM_001079909.2 | NP_001073378.1 | 15 | 2642 | |||
NM_010206.3 | NP_034336.2 | 16 | 2909 | |||
GenBank mRNA | AF176552.1 | - | 14 | 1940 | ||
AK028354.1 | - | 15 | 2082 | |||
AK034295.1 | - | 6 | 729 | |||
AK049704.1 | - | 16 | 2869 | |||
AK167639.1 | - | 7 | 2206 | |||
BC010200.1 | - | 16 | 2802 | 3967874 | ||
BC033447.1 | - | 14 | 1905 | 4037899 | ||
BC053921.1 | - | 9 | 2149 | |||
M28998.1 | - | 15 | 2207 | |||
M33760.1 | - | 14 | 1926 | |||
M65053.1 | - | 15 | 2180 | |||
U22324.1 | - | 15 | 2207 | |||
U23445.1 | - | 14 | 1940 | |||
X51893.1 | - | 16 | 2264 |