Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110498.1 | NP_001103968.1 | 6 | 743 | ||
NM_010509.2 | NP_034639.2 | 6 | 743 | |||
GenBank mRNA | AF013274.1 | - | 6 | 521 | ||
AF013486.1 | - | 6 | 615 | |||
AK003185.1 | - | 6 | 677 | |||
AK010952.1 | - | 6 | 694 | |||
AK139169.1 | - | 6 | 723 | |||
AK151474.1 | - | 6 | 671 | |||
AK151493.1 | - | 6 | 656 | |||
AK151777.1 | - | 6 | 671 | |||
AK152878.1 | - | 6 | 677 | |||
AK153317.1 | - | 6 | 677 | |||
AK199902.1 | - | 2 | 96 | |||
BC003743.1 | - | 5 | 438 | |||
BC071225.1 | - | 5 | 441 | 4187603 | ||
Y09813.1 | - | 6 | 498 | |||
Y09864.1 | - | 6 | 633 | |||
Y09865.1 | - | 6 | 633 |