Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010348.3 | NP_034478.1 | 3 | 1011 | ||
NM_146072.4 | NP_666184.2 | 3 | 1011 | |||
GenBank mRNA | AK042635.1 | - | 3 | 1012 | ||
AK043542.1 | - | 3 | 991 | |||
AK043737.1 | - | 3 | 975 | |||
AK044250.1 | - | 3 | 970 | |||
AK044420.1 | - | 3 | 965 | |||
AK046122.1 | - | 3 | 1018 | |||
AK048634.1 | - | 3 | 982 | |||
AK133667.1 | - | 3 | 997 | |||
BC031822.1 | - | 2 | 433 | 4501004 | ||
BC049275.1 | - | 2 | 433 | 4501004 | ||
X66118.1 | - | 3 | 997 |