Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRB10 SIRNA Assays ›
          Gene Symbol:
          GRB10
          Gene Name:
          growth factor receptor bound protein 10
          Gene Aliases:
          GRB-IR, Grb-10, IRBP, MEG1, RSS
          Chromosome Location:
          Chr.7: 50590063 - 50793462 on Build GRCh38
          UniGene ID:
          Hs.164060
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 16193

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GRB10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001001549.2 NP_001001549.1 4, 5 543
          NM_001001550.2 NP_001001550.1 4, 5 623
          NM_001001555.2 NP_001001555.1 5, 6 870
          NM_005311.4 NP_005302.3 4, 5 543
          XM_011515304.1 XP_011513606.1 7, 8 982
          XM_011515312.2 XP_011513614.1 6, 7 1270
          XM_011515316.1 XP_011513618.1 5, 6 815
          XM_017012031.1 XP_016867520.1 5, 6 1067
          XM_017012044.1 XP_016867533.1 4, 5 813
          XM_017012045.1 XP_016867534.1 4, 5 477
          XM_017012046.1 XP_016867535.1 8, 9 1829
          XM_017012063.1 XP_016867552.1 12, 13 2677
          XM_017012064.1 XP_016867553.1 11, 12 2561
          XM_017012065.1 XP_016867554.1 9, 10 2013
          XM_017012066.1 XP_016867555.1 7, 8 2372
          XM_017012067.1 XP_016867556.1 7, 8 875
          XM_017012068.1 XP_016867557.1 7, 8 1741
          GenBank mRNA AB000731.1 - 4, 5 518
          AF000017.1 - 4, 5 512
          AF001534.1 - 4, 5 543
          AJ271366.1 - 7, 8 982
          AK289511.1 - 6, 7 837
          BC024285.1 - 5, 6 647 4590330
          BX649028.1 - 4, 5 603
          D86962.1 - 6, 7 1275
          U34355.1 - 4, 5 543
          U66065.1 - 4, 5 623
          U69276.1 - 4, 5 625

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of Wnt signaling pathway
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          negative regulation of phosphorylation
          negative regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          insulin-like growth factor receptor signaling pathway
          SH3/SH2 adaptor activity
          insulin receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0