Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 7 | 954 | ||
NM_005426.2 | NP_005417.1 | 8 | 1086 | |||
XM_011544268.1 | XP_011542570.1 | 6 | 815 | |||
XM_011544269.2 | XP_011542571.1 | 4 | 473 | |||
XM_017002220.1 | XP_016857709.1 | 8 | 1095 | |||
GenBank mRNA | AJ318888.2 | - | 7 | 899 | ||
AK294432.1 | - | 7 | 806 | |||
BC040247.1 | - | 5 | 430 | 4454266 | ||
BC046150.1 | - | 7 | 897 | |||
BC058918.1 | - | 7 | 912 | 4537206 | ||
U58334.1 | - | 8 | 1031 |