Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166033.1 | NP_001159505.1 | 3 | 817 | ||
NM_172977.3 | NP_766565.2 | 2 | 711 | |||
GenBank mRNA | AK030351.1 | - | 2 | 638 | ||
AK141269.1 | - | 2 | 692 | |||
AK146401.1 | - | 3 | 773 | |||
AK146403.1 | - | 3 | 773 | |||
AK150650.1 | - | 2 | 693 | |||
AK151179.1 | - | 2 | 693 | |||
AK153102.1 | - | 2 | 693 | |||
BC061476.1 | - | 2 | 644 | |||
BC095998.1 | - | 2 | 650 | 30736671 | ||
BC117919.1 | - | 2 | 623 | 40097451 | ||
BC117920.1 | - | 2 | 621 | 40097456 |