Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194344.2 | NP_919325.2 | 7 | 1054 | ||
GenBank mRNA | AK028482.1 | - | 3 | 1150 | ||
AK150064.1 | - | 7 | 832 | |||
AK155330.1 | - | 7 | 815 | |||
AK159089.1 | - | 7 | 833 | |||
AK167283.1 | - | 7 | 755 | |||
AK170383.1 | - | 7 | 887 | |||
AK170396.1 | - | 7 | 834 | |||
AK171209.1 | - | 7 | 811 | |||
AK171242.1 | - | 7 | 811 | |||
BC024909.1 | - | 5 | 504 | |||
BC056461.1 | - | 5 | 398 |