Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009462.1 | NP_033488.1 | 5 | 1271 | ||
GenBank mRNA | AK011398.1 | - | 1 | 702 | ||
AK077453.1 | - | 5 | 1293 | |||
AK129083.1 | - | 5 | 1279 | |||
AK137015.1 | - | 5 | 1293 | |||
AK153675.1 | - | 5 | 1763 | |||
AK157970.1 | - | 5 | 1295 | |||
AK165156.1 | - | 5 | 1291 | |||
AK166243.1 | - | 5 | 1288 | |||
AK172443.1 | - | 5 | 1293 | |||
BC007134.1 | - | 5 | 1273 | 3491297 | ||
D84096.1 | - | 5 | 1271 |