Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005778.3 | NP_005769.1 | 3 | 287 | ||
NR_036627.2 | 3 | 287 | ||||
XM_006712917.1 | XP_006712980.1 | 3 | 287 | |||
XM_011533261.1 | XP_011531563.1 | 3 | 287 | |||
XM_011533262.1 | XP_011531564.1 | 3 | 287 | |||
XM_017005503.1 | XP_016860992.1 | 3 | 269 | |||
XM_017005504.1 | XP_016860993.1 | 3 | 287 | |||
XM_017005505.1 | XP_016860994.1 | 3 | 241 | |||
XM_017005506.1 | XP_016860995.1 | 2 | 165 | |||
XM_017005507.1 | XP_016860996.1 | 2 | 165 | |||
GenBank mRNA | AB208813.1 | - | 3 | 232 | ||
AB586690.1 | - | 3 | 218 | |||
AF091263.1 | - | 3 | 260 | |||
AF103802.4 | - | 3 | 238 | |||
AF107493.1 | - | 3 | 236 | |||
AK297445.1 | - | 3 | 243 | |||
AK302766.1 | - | 3 | 241 | |||
AK314032.1 | - | 3 | 242 | |||
AL701303.1 | - | 3 | 277 | |||
BC002957.1 | - | 3 | 233 | |||
DB080737.1 | - | 3 | 287 | |||
KU178505.1 | - | 2 | 112 | |||
KU178506.1 | - | 2 | 112 | |||
U23946.1 | - | 3 | 181 |