Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem106a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144830.3 | NP_659079.1 | 4, 5 | 459 | ||
GenBank mRNA | AK146019.1 | - | 4, 5 | 460 | ||
AK149650.1 | - | 3, 4 | 703 | |||
AK150285.1 | - | 4, 5 | 429 | |||
AK151842.1 | - | 4, 5 | 435 | |||
AK152741.1 | - | 4, 5 | 429 | |||
AK154217.1 | - | 4, 5 | 460 | |||
AK168437.1 | - | 4, 5 | 402 | |||
AK169081.1 | - | 4, 5 | 402 | |||
AK170900.1 | - | 4, 5 | 430 | |||
AK172282.1 | - | 4, 5 | 408 | |||
BC022145.1 | - | 3, 4 | 507 | 5101508 | ||
BC033268.1 | - | 4, 5 | 432 | 5037750 |