Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem106a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144830.3 | NP_659079.1 | 5, 6 | 605 | ||
GenBank mRNA | AK136954.1 | - | 1, 2 | 1599 | ||
AK146019.1 | - | 5, 6 | 606 | |||
AK149650.1 | - | 4, 5 | 849 | |||
AK150285.1 | - | 5, 6 | 575 | |||
AK151842.1 | - | 5, 6 | 581 | |||
AK152741.1 | - | 5, 6 | 575 | |||
AK154217.1 | - | 5, 6 | 606 | |||
AK168437.1 | - | 5, 6 | 548 | |||
AK169081.1 | - | 5, 6 | 548 | |||
AK170900.1 | - | 5, 6 | 576 | |||
AK172282.1 | - | 5, 6 | 554 | |||
BC022145.1 | - | 4, 5 | 653 | 5101508 |