Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB4.1L5 SIRNA Assays ›
          Gene Symbol:
          Epb4.1l5
          Gene Name:
          erythrocyte protein band 4.1 like 5
          Gene Aliases:
          1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
          Chromosome Location:
          Chr.1: 119545032 - 119649136 on Build GRCm38
          UniGene ID:
          Mm.253156
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 169842

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Epb4.1l5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001113416.1 NP_001106887.1 3 436
          NM_145506.4 NP_663481.2 3 436
          GenBank mRNA AK027983.1 - 3 390
          AK034676.1 - 3 358
          AK037024.1 - 3 391
          AK040354.1 - 3 404
          AK044959.1 - 3 374
          AK044996.1 - 3 382
          AK047523.1 - 3 363
          AK079301.1 - 3 387
          AK136125.1 - 3 391
          AK161141.1 - 3 387
          AK167449.1 - 3 436
          AK173198.1 - 3 370
          BC003937.1 - 3 421 3486543
          BC011476.1 - 3 370 4014009
          BY737035.1 - 3 436

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis involved in differentiation
          in utero embryonic development
          somitogenesis
          epithelial to mesenchymal transition
          neural plate morphogenesis
          positive regulation of cell-matrix adhesion
          epithelial cell morphogenesis
          apical constriction
          substrate-dependent cell migration, cell attachment to substrate
          ectoderm development
          endoderm development
          mesoderm development
          mesoderm migration involved in gastrulation
          unidimensional cell growth
          posttranscriptional regulation of gene expression
          positive regulation of epithelial cell migration
          positive regulation of epithelial to mesenchymal transition
          negative regulation of cell-cell adhesion
          actin cytoskeleton organization
          actomyosin structure organization
          negative regulation of protein binding
          positive regulation of protein binding
          somite rostral/caudal axis specification
          axial mesoderm development
          axial mesoderm morphogenesis
          paraxial mesoderm development
          embryonic foregut morphogenesis
          positive regulation of focal adhesion assembly
          regulation of establishment of protein localization
          left/right axis specification
          cellular response to transforming growth factor beta stimulus
          protein binding
          cytoskeletal protein binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0