Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290320.1 | NP_001277249.1 | 9, 10 | 1447 | ||
GenBank mRNA | BC027076.1 | - | 9, 10 | 1447 | 5361175 |