Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175021.3 | NP_778186.2 | 8 | 1718 | ||
GenBank mRNA | BC042901.1 | - | 8 | 1771 | 5132736 |