Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290740.1 | NP_001277669.1 | 4, 5 | 512 | ||
NM_030560.5 | NP_085037.2 | 5, 6 | 570 | |||
NM_172667.2 | NP_766255.1 | 4, 5 | 509 | |||
GenBank mRNA | AK045589.1 | - | 4, 5 | 341 | ||
AK051237.1 | - | 4, 5 | 337 | |||
AK077961.1 | - | 5, 6 | 398 | |||
AK088752.1 | - | 4, 5 | 340 | |||
AK089988.1 | - | 345 | ||||
AK129404.1 | - | 323 | ||||
AK132001.1 | - | 4, 5 | 453 | |||
AK143352.1 | - | 4, 5 | 342 | |||
AK149528.1 | - | 4, 5 | 336 | |||
AK166487.1 | - | 4, 5 | 345 | |||
BC003993.1 | - | 4, 5 | 322 | 3492146 | ||
BU613482.1 | - | 4, 5 | 509 |