Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153516.2 | NP_705736.1 | 4, 5 | 563 | ||
GenBank mRNA | AK031018.1 | - | 4, 5 | 512 | ||
AK047223.1 | - | 4, 5 | 565 | |||
AK088417.1 | - | 4, 5 | 578 | |||
AK132774.1 | - | 4, 5 | 572 | |||
AK143457.1 | - | 4, 5 | 576 | |||
AK148007.1 | - | 4, 5 | 582 | |||
AK165068.1 | - | 4, 5 | 556 | |||
AK168833.1 | - | 4, 5 | 553 | |||
AK178476.1 | - | 3, 4 | 338 | |||
AK178590.1 | - | 3, 4 | 336 | |||
AK179659.1 | - | 3, 4 | 335 | |||
AK186867.1 | - | 3, 4 | 337 | |||
AK187777.1 | - | 3, 4 | 336 | |||
BC027307.1 | - | 4, 5 | 453 | 4014844 | ||
BC029016.1 | - | 4, 5 | 563 | 5358785 |